Ethylenglycol VLSI, elektronische Qualität

Lieferant: VWR Chemicals
Achtung

Synonyme: Glycol, 1,2-Dihydroxyethan, 1,2-Ethandiol

50488857.320EA 190 EUR
50488857.320
Ethylenglycol VLSI, elektronische Qualität
Ethylenglycol
Formel: HOCH₂CH₂OH
Molecular Weight: 62,07 g/mol
Siedepunkt: 196…198 °C (1013 hPa)
Schmelzpunkt: –13 °C
Dichte: 1,115 g/cm³ (20 °C)
Flammpunkt: 116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002885
CAS-Nummer: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832

Bestellen

Spezifikation Testergebnisse

Assay (GC) ≥ 99.8 %
Identification (IR) Passes test
Miscibility with water Passes test
Colouration ≤ 10 APHA
Particle size(>0.50 μm)(/ml)(at filling) ≤ 2000
Resistivity (MOhm-cm) ≥ 1
Evaporation residue ≤ 5 ppm
Free acid (as CH₃COOH) ≤ 10 ppm
Free alkali (as NH₃) ≤ 3 ppm
Water ≤ 2000 ppm
Cl (Chloride) ≤ 4000 ppb
Ag (Silver) ≤ 20 ppb
Al (Aluminium) ≤ 100 ppb
As (Arsenic) ≤ 10 ppb
Au (Gold) ≤ 100 ppb
B (Boron) ≤ 10 ppb
Ba (Barium) ≤ 100 ppb
Be (Beryllium) ≤ 20 ppb
Bi (Bismuth) ≤ 100 ppb
Ca (Calcium) ≤ 100 ppb
Cd (Cadmium) ≤ 50 ppb
Co (Cobalt) ≤ 20 ppb
Cr (Chromium) ≤ 20 ppb
Cu (Copper) ≤ 20 ppb
Fe (Iron) ≤ 100 ppb
Ga (Gallium) ≤ 20 ppb
In (Indium) ≤ 20 ppb
K (Potassium) ≤ 100 ppb
Li (Lithium) ≤ 20 ppb
Mg (Magnesium) ≤ 100 ppb
Mn (Manganese) ≤ 20 ppb
Mo (Molybdenum) ≤ 50 ppb
Na (Sodium) ≤ 100 ppb
Ni (Nickel) ≤ 20 ppb
Pb (Lead) ≤ 50 ppb
Pt (Platinum) ≤ 200 ppb
Sb (Antimony) ≤ 10 ppb
Sn (Tin) ≤ 100 ppb
Sr (Strontium) ≤ 20 ppb
Ti (Titanium) ≤ 100 ppb
Tl (Thallium) ≤ 50 ppb
V (Vanadium) ≤ 50 ppb
Zn (Zinc) ≤ 100 ppb
Zr (Zirconium) ≤ 200 ppb

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR