N-Methyl-2-pyrrolidon (NMP) VLSI, elektronische Qualität

Lieferant: VWR Chemicals
Gefahr

50489118.320EA 210 EUR
50489118.320
N-Methyl-2-pyrrolidon (NMP) VLSI, elektronische Qualität
N-Methyl-2-pyrrolidon (NMP)
Formel: C₅H₉NO
Siedepunkt: 202 °C (1013 hPa)
Schmelzpunkt: –24 °C
Dichte: 1,033 g/cm³ (20 °C)
Flammpunkt: 91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003193
CAS-Nummer: 872-50-4
EINECS: 212-828-1
REACH: 01-2119472430-46
Merck Index: 13,06140

Bestellen

Spezifikation Testergebnisse

Assay (GC) ≥ 99.5 %
Identification (IR) Passes test
Colouration ≤ 50 APHA
Particle size(>0.50 μm)(/ml)(at filling) ≤ 80
Resistivity (MOhm-cm) ≥ 1
Free acid (as C₂H₅COOH) ≤ 50 ppm
Free amine (as methylamine) ≤ 20 ppm
Water ≤ 1000 ppm
Cl (Chloride) ≤ 500 ppb
PO₄ (Phosphate) ≤ 1000 ppb
SO₄ (Sulphate) ≤ 500 ppb
Ag (Silver) ≤ 10 ppb
Al (Aluminium) ≤ 50 ppb
As (Arsenic) ≤ 10 ppb
Au (Gold) ≤ 10 ppb
B (Boron) ≤ 10 ppb
Ba (Barium) ≤ 50 ppb
Be (Beryllium) ≤ 10 ppb
Bi (Bismuth) ≤ 10 ppb
Ca (Calcium) ≤ 50 ppb
Cd (Cadmium) ≤ 10 ppb
Co (Cobalt) ≤ 10 ppb
Cr (Chromium) ≤ 10 ppb
Cu (Copper) ≤ 10 ppb
Fe (Iron) ≤ 20 ppb
Ga (Gallium) ≤ 10 ppb
Ge (Germanium) ≤ 10 ppb
In (Indium) ≤ 10 ppb
K (Potassium) ≤ 50 ppb
Li (Lithium) ≤ 10 ppb
Mg (Magnesium) ≤ 20 ppb
Mn (Manganese) ≤ 10 ppb
Mo (Molybdenum) ≤ 20 ppb
Na (Sodium) ≤ 100 ppb
Ni (Nickel) ≤ 5 ppb
Pb (Lead) ≤ 10 ppb
Pt (Platinum) ≤ 5 ppb
Sb (Antimony) ≤ 5 ppb
Sn (Tin) ≤ 10 ppb
Sr (Strontium) ≤ 5 ppb
Ti (Titanium) ≤ 10 ppb
Tl (Thallium) ≤ 10 ppb
V (Vanadium) ≤ 10 ppb
Zn (Zinc) ≤ 10 ppb
Zr (Zirconium) ≤ 10 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR