2-Propanol SLSI, Electronic Grade

Lieferant: VWR Chemicals
Gefahr

Synonyme: Dimethylcarbinol, IPA, Isopropylalkohol, 2-Hydroxypropan, Isopropanol

50489128.320EA 158 EUR
50489128.320
2-Propanol SLSI, Electronic Grade
2-Propanol
Formel: (CH₃)₂CHOH
Molecular Weight: 60.1 g/mol
Siedepunkt: 82 °C (1013 hPa)
Schmelzpunkt: –89 °C
Dichte: 0.786 g/cm³ (20 °C)
Flammpunkt: 12 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011674
CAS-Nummer: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228

Bestellen

Spezifikation Testergebnisse

Assay (GC) ≥ 99.90 %
Identification by GC Conforms
Particle size (>0.50 μm)(/ml)(at filling) ≤ 100
Resistivity (MOhm-cm) ≥ 20
Water ≤ 200 ppm
Cl (Chloride) ≤ 100 ppb
PO₄ (Phosphate) ≤ 100 ppb
SO₄ (Sulphate) ≤ 100 ppb
Ag (Silver) ≤ 1.00 ppb
Al (Aluminium) ≤ 1.00 ppb
As (Arsenic) ≤ 1.00 ppb
Au (Gold) ≤ 1.00 ppb
B (Boron) ≤ 1.00 ppb
Ba (Barium) ≤ 1.00 ppb
Be (Beryllium) ≤ 1.00 ppb
Bi (Bismuth) ≤ 1.00 ppb
Ca (Calcium) ≤ 1.00 ppb
Cd (Cadmium) ≤ 1.00 ppb
Co (Cobalt) ≤ 1.00 ppb
Cr (Chromium) ≤ 1.00 ppb
Cu (Copper) ≤ 1.00 ppb
Fe (Iron) ≤ 1.00 ppb
Ga (Gallium) ≤ 1.00 ppb
Ge (Germanium) ≤ 1.00 ppb
In (Indium) ≤ 1.00 ppb
K (Potassium) ≤ 1.00 ppb
Li (Lithium) ≤ 1.00 ppb
Mg (Magnesium) ≤ 1.00 ppb
Mn (Manganese) ≤ 1.00 ppb
Mo (Molybdenum) ≤ 1.00 ppb
Na (Sodium) ≤ 2.00 ppb
Ni (Nickel) ≤ 1.00 ppb
Pb (Lead) ≤ 1.00 ppb
Pt (Platinum) ≤ 1.00 ppb
Sb (Antimony) ≤ 1.00 ppb
Sn (Tin) ≤ 1.00 ppb
Sr (Strontium) ≤ 1.00 ppb
Ti (Titanium) ≤ 1.00 ppb
Tl (Thallium) ≤ 1.00 ppb
V (Vanadium) ≤ 1.00 ppb
Zn (Zinc) ≤ 1.00 ppb
Zr (Zirconium) ≤ 1.00 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR