Salzsäure 36% VLSI, Electronic Grade
Lieferant: VWR Chemicals
|
Gefahr
|
Synonyme:
Hydrochloric acid solution, Muriatic acid
50489136.320EA
169
EUR
50489136.320
Salzsäure 36% VLSI, Electronic Grade
Salzsäure
Formel:
HCl Molecular Weight: 36.46 g/mol Siedepunkt: 110 °C (1013 hPa) Schmelzpunkt: –30 °C Dichte: 1.18 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011324 CAS-Nummer: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8 II REACH: 01-2119484862-27 |
Spezifikation Testergebnisse
Assay (acidimetric) | 36.0 - 38.0 % |
Colouration | ≤ 10 APHA |
Particle size(>0.50 μm)(/ml)(at filling) | ≤ 64 |
Free halogen (as Cl₂) | ≤ 0.5 ppm |
Non-volatile matter | ≤ 5 |
NH₄ (Ammonium) | ≤ 1000 ppb |
PO₄ (Phosphate) | ≤ 100 ppb |
SO₃ (Sulphite) | ≤ 500 ppb |
SO₄ (Sulphate) | ≤ 500 ppb |
Ag (Silver) | ≤ 10 ppb |
Al (Aluminium) | ≤ 50 ppb |
As (Arsenic) | ≤ 5 ppb |
Au (Gold) | ≤ 20 ppb |
B (Boron) | ≤ 10 ppb |
Ba (Barium) | ≤ 10 ppb |
Be (Beryllium) | ≤ 10 ppb |
Bi (Bismuth) | ≤ 20 ppb |
Ca (Calcium) | ≤ 200 ppb |
Cd (Cadmium) | ≤ 5 ppb |
Co (Cobalt) | ≤ 10 ppb |
Cr (Chromium) | ≤ 10 ppb |
Cu (Copper) | ≤ 10 ppb |
Fe (Iron) | ≤ 100 ppb |
Ga (Gallium) | ≤ 20 ppb |
Ge (Germanium) | ≤ 50 ppb |
In (Indium) | ≤ 10 ppb |
K (Potassium) | ≤ 50 ppb |
Li (Lithium) | ≤ 10 ppb |
Mg (Magnesium) | ≤ 50 ppb |
Mn (Manganese) | ≤ 10 ppb |
Mo (Molybdenum) | ≤ 10 ppb |
Na (Sodium) | ≤ 50 ppb |
Ni (Nickel) | ≤ 10 ppb |
Pb (Lead) | ≤ 10 ppb |
Pt (Platinum) | ≤ 20 ppb |
Sn (Tin) | ≤ 20 ppb |
Sr (Strontium) | ≤ 10 ppb |
Ti (Titanium) | ≤ 10 ppb |
Tl (Thallium) | ≤ 20 ppb |
V (Vanadium) | ≤ 10 ppb |
Zn (Zinc) | ≤ 50 ppb |
Zr (Zirconium) | ≤ 10 ppb |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...