p38 MAP Kinase Inhibitor VIII ≥97% (durch HPLC), Calbiochem®, Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem

Synonyme: (4-((2-Amino-4-bromophenyl)amino)-2-chlorophenyl)-(2-methylphenyl)methanone

M-Clarity™ Qualitätsstufe = MQ100

506163-5MG
506163-5MGEA 145 EUR
506163-5MG
p38 MAP Kinase Inhibitor VIII ≥97% (durch HPLC), Calbiochem®, Sigma-Aldrich®
p38 MAP Kinase Inhibitor VIII
Formel: C₂₀H₁₆BrClN₂O
Molecular Weight: 415.71 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 321351-00-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR