Supelco® TraceCERT® Organic Reference Standard, Dimethyl fumarate
Lieferant: Merck
50744-100MG
50744-100MGEA 81.1 EUR
50744-100MG
Supelco® TraceCERT® Organic Reference Standard, Dimethyl fumarate
Standards Organische Standards
  • GHS signal: GHS07, GHS09
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P261 - P264 - P271 - P273 - P280 - P302 + P352 + P312
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2c025a4bb29c08baa02b744dcf2246f3ab8e28b&vid=c2c025a4bb29c08baa02b744dcf2246f3ab8e28b&prid=web&clid=SAPDAM