Organic reference standard, Bisphenol B, analytical standard, Supelco®
Lieferant: Merck
50877-100MG
50877-100MGEA 77.6 EUR
50877-100MG
Organic reference standard, Bisphenol B, analytical standard, Supelco®
Standards Organische Standards
Bisphenol B is a congener of bisphenol A, used primarily for the manufacturing of phenolic resins and is also a potent food contaminant.
Bisphenol B may be used as a reference standard for the determination of bisphenol B in liquid food matrices and powdered infant formulae and human urine samples by using dispersive liquid-liquid microextraction (DLLME) with in situ derivatisation coupled with heart-cutting gas chromatography-mass spectrometry (GC-MS).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5b3db7a65796aa3e11cbee95d533ca66097eb1e&vid=b5b3db7a65796aa3e11cbee95d533ca66097eb1e&prid=web&clid=SAPDAM