Mesamoll®, Alkansulfonsäureester-Mischung, Selectophore™ ionophore, Supelco®
Lieferant: Merck
MQ100
50987-5ML
50987-5MLEA
116
EUR
50987-5ML
Mesamoll®, Alkansulfonsäureester-Mischung, Selectophore™ ionophore, Supelco®
Mesamoll®, Alkylsulphonic phenyl esters mixture
Mesamoll is an alkyl sulfonate plasticizer. Owing to its alkali resistance nature, it is widely used in membranes and sealants in the construction industry.
Good compatibility with a large number of polymers such as polyurethane (PU), polyvinyl chloride (PVC), natural rubber (NR), styrene-butadiene rubber (SBR), blends of styrene-butadiene rubber and butadiene rubber (SBR/BR), isobutylene-isoprene rubber (IIR), acrylonitrile-butadiene rubber (NBR) and chloroprene rubber (CR).
CAS-Nummer:
91082-17-6 |
Spezifikation Testergebnisse
Appearance (Color) | Colorless to Light Yellow |
Appearance (Form) | Liquid or Viscous Liquid |
Color (in APHA) | ≤ 350 APHA |
Refractive index N20/D | 1.498 - 1.502 |
Water (Coulometric) | ≤ 0.1 % |
Viscosity (Rotation) | 110 - 140 mPas |
Viscosity (Conditions) | Neat at 20 C (DIN 45) |
Saponification value | ≤ 200.0 |
RECOMMENDED RETEST PERIOD | 36 MONTHS |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...