VWR®, Analysensiebe, 250×55 mm, mit quadratischen Löchern
Lieferant: VWR Collection
510-1046EA 263 EUR
510-1046 510-1047 510-1048 510-1068 510-1069 510-1070 510-1071 510-1072 510-1073 510-1074 510-1050 510-1051 510-1052 510-1053 510-1054 510-1075 510-1076 510-1077 510-1078 510-1079 510-1080 510-1081 510-1082 510-1083 510-1084 510-1085 510-1049 510-1086 510-1055 510-1056 510-1057 510-1058 510-1059 510-1060 510-1061 510-1062 510-1063 510-1064 510-1065 510-1066 510-1067
VWR®, Analysensiebe, 250×55 mm, mit quadratischen Löchern
Siebe Analysensiebe
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Lieferumfang:More Product Information