VWR®, Analysensiebe, 450×70 mm, mit quadratischen Löchern
Lieferant: VWR Collection
510-1661EA 562 EUR
510-1661 510-1668 510-1667 510-1666 510-1665 510-1664 510-1662 510-1671 510-1660 510-1659 510-1658 510-1657 510-1656 510-1655 510-1663 510-1677 510-1684 510-1683 510-1682 510-1681 510-1680 510-1669 510-1678 510-1670 510-1676 510-1675 510-1674 510-1673 510-1672 510-1679
VWR®, Analysensiebe, 450×70 mm, mit quadratischen Löchern
Siebe Analysensiebe
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Lieferumfang: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.