Sieves for sonic sifter separator
Lieferant: ADVANTECH MANUFACTURING
L3-S40 L3-M125 L3-M20 L3-M35 L3-M50 L3-M70 L3-M90 L3-S60 L3-M10 L3-S270 L3-S70 L3-S30 L3-S50 57334-014 L3-S20 L3-S325 L3-S230 L3-S400
510-1322EA 0 EUR
510-1322 510-1366 510-1347 510-1351 510-1354 510-1358 510-1362 510-1325 510-2030 510-2033 510-2034 510-1321 510-1324 510-1326 510-1320 510-1331 510-1330 510-1343
Sieves for sonic sifter separator
Siebe Analysensiebe
These test sieves are serialised to meet the highest grade quality assurance systems for traceability of measurement devices.
  • Clear acrylic frame
  • Electroformed nickel mesh or stainless steel wire cloth
The serial number is labeled on each sieve and allows for tracing vital information such as:
- When the sieve was manufactured.
- Which lot of precision electroformed mesh or wire cloth was used to manufacture each sieve.
- Date order was shipped, including all pertinent information from your purchase order.
Standards: Comes with Certificate of Compliance for ASTM E 11 standard specifications for wire cloth or Certificate of Compliance for ASTM E 161 standard specifications for precision electroformed sieves.