VWR®, Analysensiebe, 350×60 mm
Lieferant: VWR Collection
510-3338EA 313 EUR
510-3338 510-3339 510-3340 510-3341 510-3342 510-3343 510-3349 510-3344 510-3345 510-3346 510-3347 510-3348 510-3350 510-3351 510-3352 510-3353 510-3354 510-3355 510-3357 510-3358 510-3359 510-3360 510-3361 510-3362 510-3363 510-3364 510-3356 510-3365 510-3366 510-3367 510-3368 510-3369 510-3370 510-3371 510-3372 510-3373 510-3374 510-3375 510-3376 510-3377 510-3378 510-3379 510-3380 510-3381 510-3382 510-3383 510-3384 510-3385 510-3386 510-3387 510-3388 510-3389 510-3390 510-3391 510-3392 510-3393 510-3394 510-3395 510-3396 510-3397 510-3398 510-3399 510-3400 510-3401 510-3402 510-3403 510-3404 510-3405 510-3406 510-3407 510-3408 510-3409 510-3410 510-3411 510-3412 510-3413 510-3414 510-3415 510-3416 510-3417 510-3418 510-3419 510-3420 510-3421 510-3422 510-3423 510-3424 510-3425 510-3426 510-3427 510-3428 510-3429
VWR®, Analysensiebe, 350×60 mm
Siebe Analysensiebe
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Lieferumfang: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.