Vibratory sieve shakers, Analysette®3 spartan
Lieferant: FRITSCH
03.8020.00
510-4102EA 2494 EUR
510-4102
Vibratory sieve shakers, Analysette®3 spartan
Schüttler Siebmaschinen
Analysette®3 spartanis a vibratory sieve shaker for all typical sieving tasks in the laboratory, with optical adjustment of the amplitude on the running instrument. The Analysette®3 spartan is also suitable for quantitative particle size analysis of solids and suspensions of all kinds through dry or wet sieving with woven test sieves. It is convertible into a micro mill for fine comminution of dry laboratory samples or solids in suspensions and for homogenisation of emulsions or pastes.
  • Automatic sieve analysis with sieve evaluation software autosieve
  • Quick conversion to vibratory micro mill pulverisette
  • Exact entry of the sieving time via a precise digital timer
  • Manual adjustment of the amplitude during sieving
  • Display of the amplitude during sieving
  • Ergonomically installed robust soft touchpad
  • Optimum power consumption prevents warming up of the sieving system
Up to 10 test sieves can be used simultaneously per working cycle and this system also allows up to 5 sieving operations to be performed at the same time.
Bestellinformation: Supplied with Easytwist sieve stack tensioner with steel-reinforced plastic bands and autosieve software. Clamping lid, test sieves or sieve pan and other accessories need to be ordered separately.