BCECF AM ester 1 mg/mL in anhydrischem DMSO ≥95% (durch TLC) fluoreszierender pH-Indikator

Lieferant: Biotium

51009
51009.EA 217 EUR
51009.
BCECF AM ester 1 mg/mL in anhydrischem DMSO ≥95% (durch TLC) fluoreszierender pH-Indikator
BCECF AM ester

BCECF, AM ester is membrane-permeant and thus can be loaded into cells via incubation. This product is a mixture of three molecular species, all of which are readily hydrolyzed by endogenous esterases into a single BCECF free acid form once they are in the cells.


  • Cell permeability: Membrane permeant
  • Colors: Green
  • Absorption/Emission λabs <300nm (before hydrolysis)
  • λabs\λem = 503/528 nm (pH 9, after hydrolysis)
  • Store at -20°C and protect from light and moisture

Formel: C₄₂H₄₀O₂₁
Lagertemperatur: Gefrierschrank
MDL: MFCD00036969
CAS-Nummer: 117464-70-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=825a7550c456e5d203606af9dc6266ba39c0ff67&vid=9a4aca4f3bc9c03b7d450afef197f168ffbde83c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR