BCECF (2′,7′ –bis-(Carboxyethyl)-5-(and-6)-carboxyfluorescein) ≥95% (durch HPLC) fluoreszierender pH-Indikator

Lieferant: Biotium

Synonyme: BCEF acid

51010
51010.EA 160 EUR
51010.
BCECF (2′,7′ –bis-(Carboxyethyl)-5-(and-6)-carboxyfluorescein) ≥95% (durch HPLC) fluoreszierender pH-Indikator
BCECF (2′,7′ –bis-(Carboxyethyl)-5-(and-6)-carboxyfluorescein)

BCECF (2′,7′ –bis-(Carboxyethyl)-5-(and-6)-carboxyfluorescein) is the most widely used fluorescent pH sensor. Because its pKa (6.97) is close to physiological pH, BCECF can detect cytosolic pH change with high sensitivity.


  • λEx/λEm = 508 nm/530 nm (high pH)
  • Cell permeability: Membrane impermeant
  • pKa 6.97
  • Colors: Green
  • Yellow orange solid soluble in DMSO
  • Store at 4°C and protect from light, especially in solution


At low pH, the dye is weakly fluorescent but becomes more fluorescent with increasing pH. The excitation spectrum of the dye undergoes a slight shift during pH change, while the wavelength of the emission maximum remains unchanged. The pH is determined ratiometrically by the relative fluorescent intensities at 535 nm when the dye is excited at 439 nm and 505 nm respectively. BCECF, acid is membrane-impermeant and therefore can be loaded into cells via microinjection or scrape loading. Also see the membrane permeant AM ester form of BCECF (51011).

Formel: C₂₇H₂₀O₁₁
Molecular Weight: 520,44 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00036970
CAS-Nummer: 85138-49-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d13f72d93dc4cc923d1941fa813ab59884becc7&vid=c85de5b6c0be798861b3c7db0b25e5a3f034d0c0&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR