5(6)-Carboxyfluorescein diacetate ≥95% (durch TLC) fluoreszierender pH-Indikator

Lieferant: Biotium

Synonyme: CFDA, 5(6)-Carboxy-di-O-acetylfluorescein, 5(6)-CFDA

51014
51014.EA 156 EUR
51014.
5(6)-Carboxyfluorescein diacetate ≥95% (durch TLC) fluoreszierender pH-Indikator
5(6)-Carboxyfluorescein diacetate

5(6)-CFDA (full name: 5-(and-6)-Carboxyfluorescein diacetate) is membrane-permeant and thus can be loaded into cells via incubation.


  • Cell permeability: Membrane permeant
  • Cellular localization: Nucleus & cytoplasm
  • Colors: Green
  • λex\λem 492nm\517nm (pH 9, after hydrolysis)
  • Extinction Coefficient (ε) 78,000 (pH 9, after hydrolysis)
  • Off-white solid soluble in DMSO
  • Store desiccated at 4°C


Once inside cells, CFDA is hydrolyzed by intracellular esterases to carboxyfluorescein (see catalog number 51013).

Formel: C₂₅H₁₆O₉
Molecular Weight: 460,39 g/mol
MDL: MFCD00467494
CAS-Nummer: 124387-19-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f38b3e063d3e37bfbebdc5603b56be4e07706e0&vid=cf034183f44d9e0d3fdf4a4761be643a04dfcd89&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR