5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate Gemisch aus Isomeren ≥98% (durch TLC) fluoreszierender pH-Indikator
Lieferant: Biotium
|
Achtung
|
Synonyme:
Carboxy-DCFDA, 5-(6)-Carboxydichlorofluorescein diacetate, 5(6)-CDCFDA, CDCFDA
51016
51016.EA
219
EUR
51016.
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate Gemisch aus Isomeren ≥98% (durch TLC) fluoreszierender pH-Indikator
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate
5-(and-6)-Carboxy-2′,7′-dichlorofluorescein diacetate is membrane-permeant and thus can be loaded into cells via incubation. The diacetate is readily hydrolyzed to 5-(and-6)-carboxy-2′,7′-dichlorofluorescein (51015) by intracellular esterases.
- Cell permeability: Membrane impermeant
- Cellular localization: Fluid phase tracer
- pKa 4.8
- Colors: Green
- White solid soluble in DMSO
- Store at 4°C and protect from light, especially in solution
Formel:
C₂₅H₁₄Cl₂O₉ Molecular Weight: 529,28 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08459585 CAS-Nummer: 127770-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...