Conical sleeves for vacuum filtration, Sterilin®
Lieferant: Sterilin
CON0006
511-0098EA 68.1 EUR
511-0098
Conical sleeves for vacuum filtration, Sterilin®
Halterungen und Träger Stützmanschetten
Red natural rubber cones for use with filtering flasks.
  • Set of six pieces
  • Set includes each one of 7/15 mm, 10/22 mm, 15/28 mm, 16/36 mm, 22/42 mm and 26/52 mm (inner diamater at low end/inner diameter at top end)