Vacuum-type glass membrane holder, glass reservoir, Whatman™
Lieferant: Whatman products (Cytiva)
1961-054
511-0255EA 166 EUR
511-0255
Vacuum-type glass membrane holder, glass reservoir, Whatman™
Filterhalter
Produced from borosilicate glass and available with a choice of support screen.
  • Suitable for aqueous and organic solvent filtration
The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane. The sintered glass support is recommended for filtration and biological analysis. The 304-stainless steel support screen is suitable for use with proteinaceous solutions.