Scheidetrichter, birnenförmig
Lieferant: SciLabware
2180/02M 2180/04M 2180/06M 2180/08M 2180/10M 2180/12M 2180/14M
511-0037EA 71.2 EUR
511-0037 511-0754 511-0755 511-0756 511-0757 511-0758 511-0759
Scheidetrichter, birnenförmig
Trichter Scheidetrichter
PYREX®, borosilicate glass.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Available with ground glass stopper and interchangeable PTFE key.