Büchner-Trichter, Hartley Typ
Lieferant: DIXON SCIENTIFIC
237091601 237091602 237091603
511-0837EA 423 EUR
511-0837 511-0838 511-0839
Büchner-Trichter, Hartley Typ
Trichter Filtertrichter Büchner-Trichter
Top and base: Borosilicate glass with support plate.
  • Available with choice of perforated disc material
  • Complete with three PE coated clips
  • Available for 4 filter sizes
Three piece construction.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable