Centrifugal filtration units, Vivaspin® Endotest
Lieferant: Sartorius
VS15RXETO
512-0640EA 221 EUR
512-0640
Centrifugal filtration units, Vivaspin® Endotest
Filter Zentrifugaleinheiten
For 2 to 15 ml, cellulose triacetate membrane. Disposable ultrafiltration device for endotoxin concentration and removal of substances interfering with the LAL test from liquid samples. The device is a spin column based ultrafiltration unit. It can effectively be used in swing bucket or fixed angle rotors accepting 50 ml conical bottom centrifuge tubes.
  • Minimises hands on time
  • Ideal for concentration of samples for endotoxin free applications