Zentrifugaleinheiten, Vivaspin® 15R
Lieferant: Sartorius
VS15RH22 VS15RH21 VS15RH12 VS15RH11 VS15RH02 VS15RH01 VS15RH91 VS15RH92
512-4124EA 596 EUR
512-4124 512-4123 512-4120 512-4119 512-4122 512-4121 512-4125 512-4126
Zentrifugaleinheiten, Vivaspin® 15R
Filter Zentrifugaleinheiten
Vivaspin® 15R is targeting the volume segment of 4 to 15 ml with the Hydrosart® regenerated cellulose membrane. This membrane is ideal when extremely high recovery rate and very low adsorption is required, for example in applications such as concentration and desalting of immunoglobulin fractions.
  • Highest recovery at low absorption (95 to 98%)
  • Extremely short concentration time (30-fold in 15 min.)
  • Very small holdup volume (<20 µl)
  • Crystal clear housing and volume graduations
  • Vertical twin membrane for ultra-fast concentration
  • Safe sample concentration due to dead-stop volume
Vivaspin® 15R has all the unique features of the Sartorius centrifugal concentrators, including a patented vertical twin membrane and a dead stop.