Membrane disc filters, wwPTFE (water wettable PTFE)
Lieferant: Cytiva (Formerly Pall Lab)
60538 60539 60542 60547 60548 60550 60551
513-0158EA 145 EUR
513-0158 514-1109 513-0159 513-0160 514-1110 513-0161 513-0162
Membrane disc filters, wwPTFE (water wettable PTFE)
Filter Membranfilter
The next generation of universal hydrophilic PTFE membrane with maximum chemical compatibility when filtering both aqueous solutions and aggressive solvents.
  • Hydrophilic
  • Low protein binding
HPLC certified for low extractables assuring that the filter will not add artifacts or contamination to your analysis. Low protein binding of wwPTFE membrane provides high recovery of critical proteinaceous samples.