Bottle top vacuum filtration systems, cellulose nitrate membrane, Nalgene®
Lieferant: Thermo Fisher Scientific
140-4045
513-1402EA 132 EUR
513-1402
Bottle top vacuum filtration systems, cellulose nitrate membrane, Nalgene®
Filter Flaschenaufsatzfilter
These filters are particularly useful for large volume microbiological QC samples.
  • Convenient snap-away design
  • Compatible with filter manifolds and filtering flasks
Standards: Membrane conforms to APHA standard methods, 16th edition, section 909A, B, C. Meets EPA requirements for water quality work.
Lieferumfang: Upper chamber with reference mark at 150 ml; PS cover; CN membrane; support pad; PS support plate/funnel that fits a stopper or adapter with ⁹/₁₆" hole; quick-disconnect adapter for ¹/₄ or ⁵/₁₆" tubing. Each pack of 12 funnels includes one vacuum gasket that replaces stopper or adapter with ⁹/₁₆" or larger opening.
Verpackung: Gamma sterilised, individually sealed in plastic bag.