Vacuum filtration manifold, Nalgene®
Lieferant: Thermo Fisher Scientific
DS0345-0001
513-3451EA 2520 EUR
513-3451
Vacuum filtration manifold, Nalgene®
Verteiler
Vacuum filter funnel manifold for use with Nalgene analytical filter funnels and test filter funnels.
  • Made from stainless steel
  • Two-way valve with vent port