Vakuum-Filtrationsgeräte, Rapid-Flow™ Filter MF75™, Nalgene®
Lieferant: Thermo Fisher Scientific
165-0045 166-0045 167-0045 168-0045 169-0045 524-0020 565-0020 566-0020 567-0020 568-0020 569-0020 595-3320 595-4520 596-3320 596-4520 597-3320 597-4520 565-0010 566-0010 568-0010 567-0010
734-5033EA 86.8 EUR
734-5033 734-5035 734-5037 734-5039 734-5041 514-0023 514-0024 514-0025 514-0026 514-0027 514-0028 514-0029 514-0030 514-0031 514-0032 514-0033 514-0034 514-0612 514-0613 514-0614 514-0378
Vakuum-Filtrationsgeräte, Rapid-Flow™ Filter MF75™, Nalgene®
Filter Flaschenaufsatzfilter
Hydrophilic PES membrane, PS graduated housing. Ideal for biological and pharmaceutical sterilisation requirements. Universal tissue culture membrane with lowest protein binding and extractables. Rapid-Flow™ filters with 0,2 µm PES membranes are STEM CELL TESTED.
  • 0,1 µm filters guard against mycoplasma contamination
  • 0,2 µm filters remove all bacteria, ideal for sterilisation
  • 0,45 µm filters for particle removal and fluid clarification
  • 90 mm diameter filters provide large surface area for serum and other hard to filter solutions
The new multi-column design of the membrane support delivers uniformity and consistency. The Rapid-Flow™ membrane support coupled with the PES membranes offers fast flow rates and higher throughput.