Microbiological filtration system, MBS II, Whatman™
Lieferant: Whatman products (Cytiva)
10445901
514-0054EA 193 EUR
514-0054
Microbiological filtration system, MBS II, Whatman™
Filter Membranfiltergeräte
The microbiological filtration system is used for optimal microbiological control using membranes. The design of the system, which consists of an electrical membrane dispenser, a funnel dispenser, and a vacuum manifold, leads to more reproducible results.
  • Shorter preparation time
  • High reproducibility
  • Large funnel capacity for foaming liquids
  • Funnels can be autoclaved up to 50 times
The special sealing technique ensures easy handling and a good integrity of the funnel and membrane during filtration. This reduces any cross contamination to a minimum.