Vacuum filtration units, MV 050 series, Whatman™
Lieferant: Whatman products (Cytiva)
10440020
514-0144EA 1420 EUR
514-0144
Vacuum filtration units, MV 050 series, Whatman™
Trichter Membranfiltertrichter
Stainless steel vacuum filtration devices especially suitable for microbiological applications and also for analysis of residues, contamination tests and precipitate analysis.
  • Use up to 200 °C
  • Can be sterilised by dry heat up to 180 °C