Millex® Syringe Filters, HPF, Durapore (PVDF) Membrane with Glass Fibre Pre-filter, Merck
Lieferant: Merck
SLHVM25NS SLHVMZ5NZ SLHVM25NK
514-0444EA 247 EUR
514-0444 514-0445 MILFSLHVM25NK
Millex® Syringe Filters, HPF, Durapore (PVDF) Membrane with Glass Fibre Pre-filter, Merck
Filter Spritzenvorsatzfilter
These filters include multiple media: a graduated prefilter and a membrane filter for high throughput filtration of viscous or particle-laden solutions.
  • High-density polyethylene housing compatible with the most commonly used solvents
  • Multi-layer filter can improve throughput of high particulate/viscous samples
Applications include analytical sample preparation, wine analysis and general particulate removal.