Centrifugal devices, HPLC and UHPLC grade, Nanosep® MF
Lieferant: Cytiva (Formerly Pall Lab)
ODPTFE04C34 ODPTFE04C35 ODPTFE02C34 ODPTFE02C35
514-1185EA 431 EUR
514-1185 514-1186 514-1183 514-1184
Centrifugal devices, HPLC and UHPLC grade, Nanosep® MF
Filter Zentrifugaleinheiten
These centrifugal devices are suitable for rapid sample prepration in many applications.
  • Maximum chemical compatibility for nearly all sample types
  • Extremely low extractables for reliable data
  • Low protein binding
  • Hydrophilic membranes
  • Devices can be spun at 14000×g with leak-free operation
The Nanosep® wwPTFE (hydrophilic) membrane is available in 0,2 and 0,45 µm pore sizes for HPLC and UHPLC critical samples. This next generation universally compatible filter membrane is suitable for use with nearly all sample types. It is low protein and API bindingMore Product Information