Mixed cellulose ester membranes, non-sterile, 0,2 µm, Whatman™
Lieferant: Whatman products (Cytiva)
10401706 10401712 10401714 10401726 10401731 10401721
514-2161EA 116 EUR
514-2161 514-2162 514-2163 514-0764 514-0765 514-0945
Mixed cellulose ester membranes, non-sterile, 0,2 µm, Whatman™
Filter Membranfilter
Whatman non-sterile mixed cellulose ester (MCE) filters from Cytiva are suitable for E. coli culturing, enumeration of bacteria, and other culturing and cell counting methods.
  • Plain and gridded options for ease of colony and cell counting methods
  • Supports visual enumeration of bacteria
  • Appropriate for bacterial culturing, including E. coli culturing
  • Biologically inert with good thermal stability
  • Uniform microporous structure results in high flow rates when used for filtering
Whatman MCE filters from Cytiva business are composed of cellulose acetate and cellulose nitrate, giving the membrane a more uniform surface than pure nitrocellulose filters. Browse Cytiva's other MCE filter options to find the right filter for your application.
MCE filters support microbiology applications such as E. coli culturing. The porous membrane structure enables the uptake of growth media to the bacterial colonies without chemically inhibiting their growth. The gridded surface simplifies visual enumeration of bacteria, including E. coli, enterococci, clostridia, fecal coliforms, staphylococci, and Pseudomonas aeruginosa.
Plain and gridded MCE membrane options are available. The gridded option has grid lines spaced at 3,1 mm intervals. Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity.