Filterhalter SolVac®
Lieferant: Cytiva (Formerly Pall Lab)
4020
514-4001EA 516 EUR
514-4001
Filterhalter SolVac®
Filterhalter
Simplifies clean-up and degassing of mobile phase solvents and other solutions. The versatile design fits most HPLC bottles, flasks, and containers, and eliminates the added steps of washing flasks and transferring mobile phase solvent from flask to reservoir.
  • Draws directly from HPLC solvent bottle. Less likely to spill aggressive solvents than glass funnels or disposable cups
  • Patented magnetic seal is reliable and leak proof. Eliminates the possibility of membrane shifting or tearing which can occur with aluminium clamps or threaded holders
Reusable, chemically resistant PP construction is resistant to common HPLC mobile phase solvents such as methanol, acetonitrile, and tetrahydrofuran.
Applications: Remove contaminating particulate from mobile phase or other solutions, de-gas mobile phase solvents and solutions, eliminate pour-and-wait filtration, remove microbial growth every 24 hours from aqueous buffer mobile phases.
Lieferumfang: Lieferung mit Ansaugschlauch (610 mm), Klemme, Sinker, Vakuumanschlussadapter, zwei Membran-Dichtungsmanschetten und zwei Dichtungen.