Spritzenvorsatzfilter, Acrodisc® Versapor®
Lieferant: Cytiva (Formerly Pall Lab)
4189 4190 4199 4487 4489 AP-4568 AP-4189 AP-4000 AP-4001 AP-4002 4488 4473
514-4002EA 196 EUR
514-4002 514-4104 514-4106 514-4024 514-4026 514-0482 514-0470 516-1000 516-1001 516-1002 PALL4488 PALL4473
Spritzenvorsatzfilter, Acrodisc® Versapor®
Filter Spritzenvorsatzfilter
Nützlich für die Vorfiltration von partikelbeladenen wässrigen Proben, für die Filtration von Serum und für Auflösungstests.
  • Increased pore size for reduced clogging
  • Maximized throughput
  • Wide range of pore sizes available
Membran: hydrophiles Acryl-Copolymer; PP-Gehäuse: 4, 13, Acrodisc® PSF 25 mm; Acryl-Gehäuse: 25 mm
The glass fiber membrane has very low extractables. Filters are made of 100% binder-free borosilicate glass fiber.
The Versapor® (acrylic copolymer) membrane filters feature large pore sizes ideal for prefiltration. These filters are color-coded by pore size for easy identification.
Polypropylene housing, female Luer-lock inlet and male slip Luer outlet.
Standards: Acrodisc PSF syringe filters are automation certified to assure smooth operation and worry-free performance 24 hours a day in automated workstations.
Verpackung: Syringe filters packed in cases of 200 are available in four packs of 50 or in an AutoPack™ format. The AutoPack™ format consists of eight tubes with 25 syringe filters stacked on top of each other, which is specifically designed for the Caliper™ robotic workstations.