Membranfilterscheiben, Metricel® Black PES
Lieferant: Cytiva (Formerly Pall Lab)
66586 66588 66585 66587
514-4224EA 140 EUR
514-4224 514-4226 514-4223 514-4225
Membranfilterscheiben, Metricel® Black PES
Filter Membranfilter
Kontrastgebende hydrophile modifizierte Membran aus schwarzem PES für die mikrobiologische Analyse.
  • Dark background provides excellent contrast for counting opaque colonies in microbiology labs
  • Exclusive dot grid pattern does not enhance or inhibit colony growth
  • Sharp contrast between black membrane and white grid line provides guidance while viewing and counting
Applications: Isolation and enumeration of yeast and mould colonies, spoilage organism monitoring and identification in food and beverage quality control laboratories.
Standards: CertifiedMore Product Information