1,4-Dimethylpyridinium p-toluenesulphonate ≥98%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

514888-10G
514888-10GEA 112 EUR
514888-10G
1,4-Dimethylpyridinium p-toluenesulphonate ≥98%, Sigma-Aldrich®
1,4-Dimethylpyridinium p-toluenesulphonate

1,4-Dimethylpyridinium p-toluenesulfonate,98%

Formel: C₁₄H₁₇NO₃S
Molecular Weight: 279,35 g/mol
Schmelzpunkt: 148…151 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00137110
CAS-Nummer: 78105-28-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a467b4e20f68782471290640022808a01e9b7c15&vid=0c1168c3cc07528cfb8d7944c9894253eaabe284&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR