Extraktionshülsen
Lieferant: Whatman products (Cytiva)
10350211 10350215 10350217 10350219 10350220 10350223 10350225 10350226 10350227 10350234 10350236 10350238 10350240 10350241 10350242 10350243 10350245 10350247 10350250 10350252 10350255 10350261 10350265 10350273 10350274 10350275 10350287 10350437 10350306 10350324 10350416
533-0000EA 107 EUR
533-0000 515-0080 533-0001 533-0002 533-0003 533-0004 533-0005 515-0131 515-8918 515-8923 533-0006 515-8927 533-0007 515-8929 533-0008 533-0009 533-0010 515-8932 515-8813 515-8934 533-0011 515-8940 515-0081 515-8948 515-8949 515-8814 515-8958 537-0033 537-0043 WHAT10350324 WHAT10350416
Extraktionshülsen
Extraktoren Extraktionshülsen
Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.