Filterkapseln, AcroPak™ 300
Lieferant: Cytiva (Formerly Pall Lab)
12082 12085
515-0143EA 223 EUR
515-0143 PALL12085
Filterkapseln, AcroPak™ 300
Filter Membranfilterkapseln
Filtration capsules for high airflow rate venting applications. Also useful for aggressive solvent filtration.
  • 100% integrity tested
PTFE hydrophobic membrane, PP housing.