Moisture test paper, Whatman™
Lieferant: Whatman products (Cytiva)
5401-090E
515-2877EA 67.8 EUR
515-2877
Moisture test paper, Whatman™
Filter Glasfaserfilter
These borosilicate glass papers are designed for use when drying samples during moisture assessment.
Max. recommended temperature: 550 °C