Qualitative Filterpapiere, Standardsorten, Sorte 591, Whatman™
Lieferant: Whatman products (Cytiva)
10311387
515-4115EA 1110 EUR
515-4115
Qualitative Filterpapiere, Standardsorten, Sorte 591, Whatman™
Filter Filterpapiere Qualitatives Filterpapier
These cellulose filters are used in qualitative analytical techniques to determine and identify materials.
  • Thick filter paper with very high loading capacity
  • High absorbency and increased wet strength
A standard grade filter for fast filtration of medium-to-coarse precipitates.