Organic reference standard, Methyl heneicosanoate, analytical standard, Supelco®
Lieferant: Merck
51535-1G
51535-1GEA 559 EUR
51535-1G
Organic reference standard, Methyl heneicosanoate, analytical standard, Supelco®
Standards Organische Standards
Methyl heneicosanoate can find applications as an internal standard in the quantitative determination of fatty acids and their isomers composition using gas chromatography method of analysis (GC).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a11a177b050255c2229e18685f84361c55fe16c&vid=5b7d6c217bed2846a0dc697ccaa404de345fa1ae&prid=web&clid=SAPDAM