Organic reference standard, Methyl heneicosanoate, analytical standard, Supelco®
Lieferant: Merck
51535-1G
51535-1GEA
559
EUR
51535-1G
Organic reference standard, Methyl heneicosanoate, analytical standard, Supelco®
Standards
Organische Standards
Methyl heneicosanoate can find applications as an internal standard in the quantitative determination of fatty acids and their isomers composition using gas chromatography method of analysis (GC).