Supelco® TraceCERT® Organic Reference Standard, ʟ-Aspartic acid
Lieferant: Merck
51572-100MG
51572-100MGEA
86.4
EUR
51572-100MG
Supelco® TraceCERT® Organic Reference Standard, ʟ-Aspartic acid
Standards
Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P264 - P270 - P301 + P312 - P501