Mehrfach-Vakuumfiltrationsleiste, Whatman™
Lieferant: Whatman products (Cytiva)
10444835 10445835 10498761 10498762
514-0218EA 8480 EUR
514-0218 516-0501 516-0418 516-0419
Mehrfach-Vakuumfiltrationsleiste, Whatman™
Verteiler
Multiple vacuum filtration apparatus suitable only for vacuum operation.
  • For microbiological quality control
  • Residue analyses
  • Serial filtration carried out rapidly and easily with a common drainage outlet
Stainless steel including sieve filter support.
Individual stopcocks, 47/50 mm filter size, tubing nozzle 9 mm vacuum connection.
Bitte beachten Suitable only for vacuum. Do not exceed 1.3bar (300mbar overpressure) when using flushing tubes.