Ultrafree®-CL Centrifugal Filter Units with Microporous Durapore® PVDF Membrane
Lieferant: Merck
UFC40GV00 UFC40HV00 UFC40VV00 UFC40VV25 UFC40GV25 UFC40DV25 UFC40SV25
MILFUFC40GV00EA 644 EUR
MILFUFC40GV00 MILFUFC40HV00 MILFUFC40VV00 MILFUFC40VV25 516-0542 516-0541 MILFUFC40SV25
Ultrafree®-CL Centrifugal Filter Units with Microporous Durapore® PVDF Membrane
Filter Zentrifugaleinheiten
These centrifugal filter units can be used for a number of applications, including removal of particles and precipitates from aqueous and some solvent-based samples. Ideal for use with protein and nucleic acid solutions.
  • Pre-sterilised units also available
  • Fast filtration and highly reproducible performance
  • Use in fixed-angle rotors for 15 ml tubes
Hydrophilic PVDF membranes and PP housing.