Acro® 500 Filter Capsule with Supor® EKV Membrane and Polycarbonate Housing
Lieferant: Cytiva (Formerly Pall Lab)
12337
516-1169EA 0 EUR
516-1169
Acro® 500 Filter Capsule with Supor® EKV Membrane and Polycarbonate Housing
Filter Membranfilterkapseln
The Acro 500 filter capsule with Supor EKV membrane and polycarbonate housing offers a 500 cm² effective filtration area (EFA) for the sterile filtration of larger volumes of laboratory solutions.
  • Membrane features a built-in dual layer polyethersulfone (PES) construction which provides fast and efficient filtration
The low protein and preservative binding properties of the membrane allow for maximum transmission of active ingredients as well as broad liquid compatibility, ideal for the filtration of a wide range of fluids including buffers, biological fluids, and tissue culture media. With an outer housing made from polycarbonate, the improved optical clarity allows for easier venting of the capsule when in use to maximize membrane area and ultimately flow rate and throughput.