Getränke-Monitore, Microcheck® II
Lieferant: Cytiva (Formerly Pall Lab)
4761 4763 4764 4761ME
516-0201EA 185 EUR
516-0201 516-0203 516-0204 516-1227
Getränke-Monitore, Microcheck® II
Trichter Membranfiltertrichter
MicroCheck™ II beverage monitor, is an easy-to-use disposable filter funnel designed to meet the needs for microbial analysis of beverages.
  • Membrane diameter: 47 mm
  • Diameter: 6,1 cm
  • Membrane size: 0,45 µm
Polypropylene construction eliminates compatibility or extractables issues. Ergonomically designed units allow for removal of funnel cylinder with a squeeze. The base and cover convert to a petri dish for culturing the membrane in place, or the membrane can be cultured on a separate petri dish. Unit fits directly onto a standard manifold, or a stopper/adapter combination can be used for other manifold styles. Gamma irradiated. Bulk packed.
External graduation marks according to standard methods for the analysis of water and waste water.