Pleated capsules with Versapor® membrane
Lieferant: Cytiva (Formerly Pall Lab)
12116 12117
516-7516EA 208 EUR
516-7516 516-7517
Pleated capsules with Versapor® membrane
Filter Membranfilterkapseln
These disposable capsules offer easy microfiltration for laboratories and small-volume processors.The pleated capsules combine the efficiency of a membrane disc filter with the large filtration capacity of a pleated surface.
  • Capsule membranes are sealed in the housing
  • Can be used inlet up, inlet down, or horizontally
  • For gravitydriven microfiltration, capsules can be placed on a benchtop or connected to plastic tubing
PC housing, PP core, acrylic copolymer media on a non-woven support, polyester filter support.