Druckfiltrationstrichter
Lieferant: Cytiva (Formerly Pall Lab)
4280
516-7607EA 2620 EUR
516-7607
Druckfiltrationstrichter
Filterhalter
This 47 mm pressure filtration funnel features a stainless steel construction that offers excellent chemical resistance. Convenient hand tightening makes filter changing easy.