Ultralab™ Systems
Lieferant: Cytiva (Formerly Pall Lab)
FS007X70
516-8200EA 5960 EUR
516-8200
Ultralab™ Systems
Filter Tangentialfluß-Systeme
Ultralab™ systems, composed of Ultrareservoir™ containers and Ultrapump™ II pumps, are designed as a rapid, efficient replacement of dialysis for buffer exchange and desalting of macromolecules.
  • Efficient replacement of dialysis
  • Airtight lid
  • Siphoning action
Systems are specifically designed for use with Ultrasette™ lab tangential flow devices (see 29300-988 series).
Ultrapump™ II has a 0 to 600 rpm rated, ¹/₁₀ hp motor with continuous-duty rated drive, and 0 to 10 dial speed control. Pump size: 16,3×34×13,7 cm (6¹³/₃₂×13¹³/₃₂×5¹³/₃₂"). Maximum pressure: 2,8 bar intermittent, 1,8 bar continuous. Min. recirculation volume: 20 to 30 ml. Ultrareservoir™ is constructed from acrylic with EPR lid O-ring, stainless steel gauge, and polypropylene valves, barb connectors, and lid.
Airtight lid and siphoning action maintain constant sample volume and eliminate the need for multiple buffer exchanges. Sample port allows maximum sample recovery without decanting. Peristaltic pump action excludes air in the system, reduces foaming, and minimizes shear of sensitive molecules.
Bestellinformation: Ultralab™ systems consist of an Ultrareservoir™ container, Ultrapump™ II pump, and Mini-Ultrasette™ Flow Device Accessory Kit (500 ml) or Ultrasette™ Flow Device Accessory Kit (2 and 5 L). Ultrareservoir™ containers include 0 to 4,2 bar pressure gauge, assorted fittings, and 3-way valve. The 5 L container connects to Ultrasette™ device only, and is supplied with assorted fittings. The Mini-Ultrasette™ kit contains 1,8 m of PharMed® #15 tubing, 45 cm of 3,2 mm Int.Ø. Tygon® filtrate tubing, eight stainless steel tubing clamps, two barb-to-Luer fittings, and one screw clamp. The Ultrasette™ kit contains 1,8 m of PharMed® #24 tubing, 60 cm of 4,8 mm Int.Ø. Tygon® filtrate tubing, eight stainless steel tubing clamps, one barb fitting, and one screw clamp.