Belüftungsfilter, Midisart® 2000
Lieferant: Sartorius
17805-TCN
517-0014EA 0 EUR
517-0014
Belüftungsfilter, Midisart® 2000
Filter Belüftungsfilter
Venting filters for the sterile venting of small fermenters and culture vessels.
Hydrophobic PP reinforced PTFE membrane, PP housing